Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fgfr-1 beta protein, human (HEK293, hFc) | 91.9% | 50UG
SDP

Catalog No. 5000177330
Encompass_Preferred

Recombinant human FGFR-1 beta (beta isoform) is supplied as an hFc fusion protein expressed in HEK293 cells and provided as a lyophilized material for use in receptor-ligand interaction studies, signaling assays, and screening applications. It is characterized by a defined molecular-weight range and quality controls appropriate for research use.

  • Expressed in HEK293 cells for human-like post-translational modifications.
  • Provided as an hFc fusion to facilitate detection and immobilization.
  • Molecular weight approximately 97.5-110 kDa.
  • Purity greater than 90% (lot example: 91.87%).
  • Lyophilized from 20 mM PB, 150 mM NaCl, pH 7.4 formulation buffer.
  • Store at -20°C for long-term storage; after reconstitution keep at 4°C short term or aliquot and freeze at -20°C/-80°C.
  • Intended for research use only.

Catalog No. 50-001-77330 Supplier Medchemexpress LLC Supplier No. HYP70043650UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant human FGFR-1 beta (beta isoform) is supplied as an hFc fusion protein expressed in HEK293 cells and provided as a lyophilized material for use in receptor-ligand interaction studies, signaling assays, and screening applications. It is characterized by a defined molecular-weight range and quality controls appropriate for research use.

  • Expressed in HEK293 cells for human-like post-translational modifications.
  • Provided as an hFc fusion to facilitate detection and immobilization.
  • Molecular weight approximately 97.5-110 kDa.
  • Purity greater than 90% (lot example: 91.87%).
  • Lyophilized from 20 mM PB, 150 mM NaCl, pH 7.4 formulation buffer.
  • Store at -20°C for long-term storage; after reconstitution keep at 4°C short term or aliquot and freeze at -20°C/-80°C.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.