Learn More
Medchemexpress LLC FGFR-1 beta (IIIb) protein, human (HEK293, Fc) | >95.0% | 10 UG
SDP
Catalog No. 5000294352
Encompass_Preferred
Recombinant human FGFR-1 beta (IIIb) Fc-tagged protein expressed in HEK293 cells. Supplied as a C-terminal human Fc-tagged dimeric protein in 0.22 μm filtered PBS (pH 7.4), purified to >95% by reducing SDS-PAGE, with a reported molecular weight of 76-106 kDa. Provided in 10 μg aliquots for research use in receptor binding, signaling studies, and assay development. Store at -80°C and avoid repeated freeze-thaw cycles.
  • Fc-tag facilitates detection and purification.
  • High purity (>95%) suitable for biochemical assays.
  • Expressed in mammalian HEK293 cells for native post-translational modifications.
  • Supplied in PBS, pH 7.4, 0.22 μm filtered, ready for downstream assays.
  • Stable when stored at -80°C; aliquot to minimize freeze-thaw.
Catalog No. 50-002-94352 Supplier Medchemexpress LLC Supplier No. HYP7576510UG
Recombinant human FGFR-1 beta (IIIb) Fc-tagged protein expressed in HEK293 cells. Supplied as a C-terminal human Fc-tagged dimeric protein in 0.22 μm filtered PBS (pH 7.4), purified to >95% by reducing SDS-PAGE, with a reported molecular weight of 76-106 kDa. Provided in 10 μg aliquots for research use in receptor binding, signaling studies, and assay development. Store at -80°C and avoid repeated freeze-thaw cycles.
  • Fc-tag facilitates detection and purification.
  • High purity (>95%) suitable for biochemical assays.
  • Expressed in mammalian HEK293 cells for native post-translational modifications.
  • Supplied in PBS, pH 7.4, 0.22 μm filtered, ready for downstream assays.
  • Stable when stored at -80°C; aliquot to minimize freeze-thaw.