Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fgfr-1 beta (IIIb) protein, human (HEK293, Fc) | >95.0% | 50 UG
SDP

Catalog No. 5000294610
Encompass_Preferred

Recombinant human FGFR-1 beta (IIIb) extracellular domain expressed in HEK293 cells, supplied with a C-terminal human Fc tag, and provided as a sterile, 0.22 μm filtered PBS solution. Characterized by reducing SDS-PAGE and reported as >95% pure. Recommended storage is -80 °C with avoidance of repeated freeze-thaw cycles.

  • Expressed in HEK293 cells
  • C-terminal human Fc tag for detection and capture
  • Purity >95% by reducing SDS-PAGE
  • Molecular weight reported as 76-106 kDa
  • Supplied in PBS, pH 7.4, 0.22 μm filtered
  • Available sizes include 10 μg, 50 μg, and 100 μg
  • Store at -80 °C; avoid repeated freeze-thaw
  • Suitable for biochemical and cell-based FGFR studies

Catalog No. 50-002-94610 Supplier Medchemexpress LLC Supplier No. HYP7576550UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human FGFR-1 beta (IIIb) extracellular domain expressed in HEK293 cells, supplied with a C-terminal human Fc tag, and provided as a sterile, 0.22 μm filtered PBS solution. Characterized by reducing SDS-PAGE and reported as >95% pure. Recommended storage is -80 °C with avoidance of repeated freeze-thaw cycles.

  • Expressed in HEK293 cells
  • C-terminal human Fc tag for detection and capture
  • Purity >95% by reducing SDS-PAGE
  • Molecular weight reported as 76-106 kDa
  • Supplied in PBS, pH 7.4, 0.22 μm filtered
  • Available sizes include 10 μg, 50 μg, and 100 μg
  • Store at -80 °C; avoid repeated freeze-thaw
  • Suitable for biochemical and cell-based FGFR studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.