Learn More
Medchemexpress LLC FGFR-1 beta IIIc 100ug | 100ug
SDP
Catalog No. 5000187294
Encompass_Preferred
FGFR-1 alpha is a tyrosine-protein kinase receptor for fibroblast growth factor that is critical in embryonic development cell proliferation differentiation and migration GnRH is essential for mesodermal patterning axial organization skeletogenesis and nervous system development FGFR-1 beta (IIIc) Protein Human (HEK293 His-Avi) is the recombinant human-derived FGFR-1 beta expressed by HEK293 with C-Avi C-His labeled tag
Catalog No. 50-001-87294 Supplier Medchemexpress LLC Supplier No. HYP77654100UG
FGFR-1 alpha is a tyrosine-protein kinase receptor for fibroblast growth factor that is critical in embryonic development cell proliferation differentiation and migration GnRH is essential for mesodermal patterning axial organization skeletogenesis and nervous system development FGFR-1 beta (IIIc) Protein Human (HEK293 His-Avi) is the recombinant human-derived FGFR-1 beta expressed by HEK293 with C-Avi C-His labeled tag