Learn More
Medchemexpress LLC FGFR-1 beta IIIc 20ug | 20ug
SDP
Catalog No. 5000187324
Encompass_Preferred
FGFR-1 alpha is a tyrosine-protein kinase receptor for fibroblast growth factor that is critical in embryonic development cell proliferation differentiation and migration GnRH is essential for mesodermal patterning axial organization skeletogenesis and nervous system development FGFR-1 beta (IIIc) Protein Human (HEK293 His-Avi) is the recombinant human-derived FGFR-1 beta expressed by HEK293 with C-Avi C-His labeled tag
Catalog No. 50-001-87324 Supplier Medchemexpress LLC Supplier No. HYP7765420UG
FGFR-1 alpha is a tyrosine-protein kinase receptor for fibroblast growth factor that is critical in embryonic development cell proliferation differentiation and migration GnRH is essential for mesodermal patterning axial organization skeletogenesis and nervous system development FGFR-1 beta (IIIc) Protein Human (HEK293 His-Avi) is the recombinant human-derived FGFR-1 beta expressed by HEK293 with C-Avi C-His labeled tag