Learn More
Medchemexpress LLC FGFR-3 Human HEK29 2ug
SDP
Catalog No. 5000196884
Encompass_Preferred
Recombinant human FGFR-3 alpha (IIIc) extracellular domain fused to a human Fc, expressed in HEK293 cells and supplied as a lyophilized powder for research use. The protein is validated for bioactivity and quality, with low endotoxin and high purity to support biochemical and cell-based assays.
  • Human extracellular FGFR-3 domain fused to human Fc for improved stability and detection.
  • Expressed in HEK293 cells to provide mammalian post-translational modifications.
  • Lyophilized formulation for long-term storage and transport.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Endotoxin below 1 EU/μg as measured by LAL.
  • Bioactivity validated in cell-based proliferation assays with low ng/mL ED50.
  • Reconstitute at ≥100 μg/mL; add carrier protein for long-term storage.
  • Store frozen; stable at -20°C for long-term, and 4°C short-term after reconstitution.
Catalog No. 50-001-96884 Supplier Medchemexpress LLC Supplier No. HYP708172UG
Recombinant human FGFR-3 alpha (IIIc) extracellular domain fused to a human Fc, expressed in HEK293 cells and supplied as a lyophilized powder for research use. The protein is validated for bioactivity and quality, with low endotoxin and high purity to support biochemical and cell-based assays.
  • Human extracellular FGFR-3 domain fused to human Fc for improved stability and detection.
  • Expressed in HEK293 cells to provide mammalian post-translational modifications.
  • Lyophilized formulation for long-term storage and transport.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Endotoxin below 1 EU/μg as measured by LAL.
  • Bioactivity validated in cell-based proliferation assays with low ng/mL ED50.
  • Reconstitute at ≥100 μg/mL; add carrier protein for long-term storage.
  • Store frozen; stable at -20°C for long-term, and 4°C short-term after reconstitution.