Learn More
Eurofins Discoverx FGFR1, active, 10 µg. (14-582)
Supplier:  Eurofins Discoverx 14582
Encompass
N-terminal GST-tagged recombinant, human FGFR1 amino acids 456-765. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9003