Learn More
Genscript Corporation FGFR1 alpha IIIcBiotin Hi
Supplier:  Genscript Corporation Z04083500
Encompass
Fibroblast growth factors (FGFs) comprise a family of at least eighteen structurally related proteins that are involved in a multitude of physiological and pathological cellular processes including cell growth differentiation angiogenesis wound healing and tumorgenesis The biological activities of the FGFs are mediated by a family of type I transmembrane tyrosine kinases which undergo dimerization and autophosphorylation after ligand binding FGFR1 is tyrosine-protein kinase that acts as cell-surface receptor for fibroblast growth factors and plays an essential role in the regulation of embryonic development cell proliferation differentiation and migration Required for normal mesoderm patterning and correct axial organization during embryonic development normal skeletogenesis and normal development of the gonadotropin-releasing hormone (GnRH) neuronal system
Catalog No. 50-000-57402