Learn More
Selleck Chemical LLC FGFR1/DDR2 inhibitor 1
SDP
Supplier:  Selleck Chemical LLC E06215MG
Encompass_Preferred
FGFR1/DDR2 inhibitor 1 is an inhibitor of fibroblast growth factor receptor 1 (FGFR1) and discoidin domain receptor 2 (DDR2)
Catalog No. 50-313-6433