Learn More
Medchemexpress LLC HY-114311 5mg Medchemexpress, FGFR1/DDR2 inhibitor 1 CAS:2308497-58-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1143115MG
Encompass_Preferred
Medchemexpress, HY-114311 5mg FGFR1/DDR2 inhibitor 1 CAS:2308497-58-5 FGFR1/DDR2 inhibitor 1 is an orally active inhibitor of fibroblast growth factor receptor 1 (FGFR1) and discoindin domain receptor 2 (DDR2), with IC50 values of 31.1 nM and 3.2 nM, respectively. Antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0869