Learn More
Medchemexpress LLC FGFR1/DDR2 inhibitor 10mM 1mL | 2308497-58-5 | 1 ML
SDP
Catalog No. 5000229405
Encompass_Preferred
FGFR1/DDR2 inhibitor 1 is a small-molecule kinase inhibitor used in preclinical research to inhibit FGFR1 and DDR2. It is supplied as a 10 mM solution in DMSO or as solid quantities for biochemical and cellular assays.
  • Potent inhibition of FGFR1 and DDR2
  • High purity (98.26%)
  • Molecular weight 501.50 g/mol
  • Available as 10 mM solution in DMSO or in solid quantities
  • Suitable for kinase assays and target validation studies
  • Store powder and solutions under recommended conditions to maintain stability
Catalog No. 50-002-29405 Supplier Medchemexpress LLC Supplier No. HY11431110MM 1ML
Add to Cart
Add to Cart
FGFR1/DDR2 inhibitor 1 is a small-molecule kinase inhibitor used in preclinical research to inhibit FGFR1 and DDR2. It is supplied as a 10 mM solution in DMSO or as solid quantities for biochemical and cellular assays.
  • Potent inhibition of FGFR1 and DDR2
  • High purity (98.26%)
  • Molecular weight 501.50 g/mol
  • Available as 10 mM solution in DMSO or in solid quantities
  • Suitable for kinase assays and target validation studies
  • Store powder and solutions under recommended conditions to maintain stability