Learn More
ACROBiosystems ACROBiosystems Human FGF R1 / CD331 Protein, His Tag
Supplier:  ACROBiosystems FG1H5223200UG
Encompass_Preferred
Recombinant Protein;200UG;Human FGF R1, His Tag (FG1-H5223) is expressed from human 293 cells (HEK293). It contains AA Arg 22 - Ile 376 (Accession # P11362-7).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 41.2 kDa. The protein migrates as 60-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human FGF basic, Tag Free (Cat. No. BFF-H4117) at 5 μg/mL (100 μL/well) can bind Human FGF R1, His Tag (Cat. No. FG1-H5223) with a linear range of 0.156-1.25 μg/mL (QC tested).
Catalog No. 50-195-6482