Learn More
Sino Biological Recombinant Human IRE1 Protein (R727A, GST Tag), Active
Catalog No. 502575008
Encompass_Preferred
Recombinant human ERN1 (R727A) (468-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5008 Supplier Sino Biological Supplier No. E3112BG500
Recombinant human ERN1 (R727A) (468-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.