Learn More
Genscript Corporation FGFR2 alpha IIIc His Human
Catalog No. 5000056927
Encompass
Four distinct genes encoding closely related FGF receptors FGF R1 - 4 are known All four genes for FGF Rs encode proteins with an N-terminal signal peptide three immunoglobulin (Ig)-like domains an acid-box region containing a run of acidic residues between the IgI and IgII domains a transmembrane domain and the split tyrosine-kinase domain Multiple forms of FGF R1 - 3 are generated by alternative splicing of the mRNAs A frequent splicing event involving FGF R1 and 2 results in receptors containing all three Ig domains referred to as the alpha isoform or only IgII and IgIII referred to as the beta isoform
Catalog No. 50-000-56927 Supplier Genscript Corporation Supplier No. Z04256100
Four distinct genes encoding closely related FGF receptors FGF R1 - 4 are known All four genes for FGF Rs encode proteins with an N-terminal signal peptide three immunoglobulin (Ig)-like domains an acid-box region containing a run of acidic residues between the IgI and IgII domains a transmembrane domain and the split tyrosine-kinase domain Multiple forms of FGF R1 - 3 are generated by alternative splicing of the mRNAs A frequent splicing event involving FGF R1 and 2 results in receptors containing all three Ig domains referred to as the alpha isoform or only IgII and IgIII referred to as the beta isoform