Learn More
Aviva Systems Biology Corp FGFR2 Antibody - middle region (ARP60045_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60045_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FGFR2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human FGFR2. Alternate names: BEK, BFR-1, CD332, CEK3, CFD1, ECT1, FLJ98662, JWS, K-SAM, KGFR, TK14, TK25ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-286-931