Learn More
Sino Biological Recombinant Human FGFR2 Protein (C491S, GST Tag), Active
Catalog No. 502574932
Encompass_Preferred
Recombinant human FGFR2 (C491S) (285-end) was expressed by baculovirus in Sf9 cells using an N-terminal GST tag.
Catalog No. 50-257-4932 Supplier Sino Biological Supplier No. F0512JG500
Recombinant human FGFR2 (C491S) (285-end) was expressed by baculovirus in Sf9 cells using an N-terminal GST tag.