Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FGFR2-IN-2 | 2677709-81-6 | 98.3% | 370.45 | C23H22N4O | 5 MG
SDP

Catalog No. 5000221057
Encompass_Preferred

FGFR2-IN-2 (Compound 38) is a small-molecule research reagent that selectively inhibits fibroblast growth factor receptor 2 (FGFR2). It exhibits potent biochemical activity against FGFR2 with reduced activity versus FGFR1 and FGFR3, and is supplied as a powder for laboratory reconstitution and storage.

  • Selective FGFR2 inhibition with IC50 of 29 nM.
  • Differential activity for FGFR1 and FGFR3 (389 nM and 758 nM) supports isoform selectivity studies.
  • Supplied as a powder for convenient reconstitution and long-term storage.
  • Storage stability: powder stable at -20°C for 3 years and 4°C for 2 years.
  • Molecular formula C23H22N4O and molecular weight 370.45 for dosing and calculations.

Catalog No. 50-002-21057 Supplier Medchemexpress LLC Supplier No. HY1452315MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FGFR2-IN-2 (Compound 38) is a small-molecule research reagent that selectively inhibits fibroblast growth factor receptor 2 (FGFR2). It exhibits potent biochemical activity against FGFR2 with reduced activity versus FGFR1 and FGFR3, and is supplied as a powder for laboratory reconstitution and storage.

  • Selective FGFR2 inhibition with IC50 of 29 nM.
  • Differential activity for FGFR1 and FGFR3 (389 nM and 758 nM) supports isoform selectivity studies.
  • Supplied as a powder for convenient reconstitution and long-term storage.
  • Storage stability: powder stable at -20°C for 3 years and 4°C for 2 years.
  • Molecular formula C23H22N4O and molecular weight 370.45 for dosing and calculations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.