Learn More
Eurofins Discoverx FGFR2 (N549H), active, 250 µg. (14-742M)
Supplier:  Eurofins Discoverx 14742M
Encompass
N-terminal 6xHis-tagged recombinant, human FGFR2 (N549H) amino acids 456-770. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9272