Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human FGFR2 [N549H], GST Tag Recombinant Protein

Catalog No. A33726
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A33726 1 mg
A33503 10 μg
A33557 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A33726 Supplier Invitrogen™ Supplier No. A33726
Add to Cart
Add to Cart
Request a Quote

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This FGFR2 N549H recombinant human protein (285-end) was expressed in insect cells. FGFR2 N549H recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Z'-LYTE™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Recombinant, Active, Cytoplasmic, Mutant The gene accession number for FGFR2 [N549H] is AAH39243.2.

FGFR2 receptor tyrosine kinase is a member of the fibroblast growth factor receptor family which plays a role in mitogenesis and differentiation. FGFR2 is a high-affinity receptor for acidic, basic and/or keratinocyte growth factor, and mutations in FGFR2 are associated with Crouzon syndrome, Pfeiffer syndrome, Craniosynostosis, Apert syndrome, Jackson-Weiss syndrome, Saethre-Chotzen syndrome, and syndromic craniosynostosis. FGFR2 is required for early postimplantation development between implantation and the formation of the egg cylinder. FGFR2 contributes to the outgrowth, differentiation, and maintenance of the inner cell mass.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P21802
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 25% glycerol, 0.25mM DTT, 0.1mM PMSF, 150mM NaCl, 10mM glutathione, 0.1mM EDTA and no preservative; pH 7.5
Gene ID (Entrez) 2263
Molecular Weight (g/mol) 74 kDa
Name Human FGFR2 [N549H], GST Tag
pH Range 7.5
Purification Method Purified
Quantity 1 mg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias FGFR2
Common Name FGFR2 (N549H Mutant)
Gene Symbol FGFR2
Kinase Family FGF Receptors
Protein Length 285-end
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 285-end
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Shipping Condition Dry Ice
Expression System Insect cells
Protein Family Growth Factors & Receptors
Protein Form Recombinant, Active, Cytoplasmic, Mutant
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.