Learn More
Sino Biological Recombinant Human FGFR1OP & FGFR1 Protein (GST Tag), Active
Supplier:  Sino Biological F0419BG500
Encompass_Preferred
Recombinant human fusion protein FGFR1OP (1-173)-FGFR1 (429-end) was expressed by baculovirus in Sf9 cells using an N-terminal GST tag.
Catalog No. 50-257-4954