Learn More
Eurofins Discoverx FGFR3, active, 10 µg. (14-464)
Supplier:  Eurofins Discoverx 14464
Encompass
N-terminal 6xHis-tagged recombinant, human FGFR amino acids 447-761. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8831