Learn More
Genscript Corporation FGFR3 beta IIIbBiotin His
Catalog No. 5000058189
Encompass
Four distinct genes encoding closely related FGF receptors FGF R1-4 are known All four genes for FGF Rs encode proteins with an N-terminal signal peptide three immunoglobulin (Ig)-like domains an acid box region containing a run of acidic residues between the IgI and IgII domains a transmembrane domain and the split tyrosine-kinase domain FGFR3 is tyrosine-protein kinase that acts as cell-surface receptor for fibroblast growth factors and plays an essential role in the regulation of cell proliferation differentiation and apoptosis
Catalog No. 50-000-58189 Supplier Genscript Corporation Supplier No. Z04090500
Four distinct genes encoding closely related FGF receptors FGF R1-4 are known All four genes for FGF Rs encode proteins with an N-terminal signal peptide three immunoglobulin (Ig)-like domains an acid box region containing a run of acidic residues between the IgI and IgII domains a transmembrane domain and the split tyrosine-kinase domain FGFR3 is tyrosine-protein kinase that acts as cell-surface receptor for fibroblast growth factors and plays an essential role in the regulation of cell proliferation differentiation and apoptosis