Learn More
Cayman Chemical FGFR3 EXrAClular DomaInh 50ug
SDP
Catalog No. 502914998
Encompass_Preferred
Source: Recombinant human C-terminal His-tagged FGFR3 extracellular domain expressed in HEK293 cells • Amino acids: 23-375 • MW: 39.6 kDa
Catalog No. 50-291-4998 Supplier Cayman Chemical Supplier No. 3702350 UG
Source: Recombinant human C-terminal His-tagged FGFR3 extracellular domain expressed in HEK293 cells • Amino acids: 23-375 • MW: 39.6 kDa