Learn More
Cayman Chemical FGFR3 Extracellular Domain (human, recombinant), Purity: ≥95% estimated by SDS-PAGE, 1 mg
SDP
Catalog No. 502915490
Encompass_Preferred
Source: Recombinant human C-terminal His-tagged FGFR3 extracellular domain expressed in HEK293 cells • Amino acids: 23-375 • MW: 39.6 kDa
Catalog No. 50-291-5490 Supplier Cayman Chemical Supplier No. 370231 MG
Source: Recombinant human C-terminal His-tagged FGFR3 extracellular domain expressed in HEK293 cells • Amino acids: 23-375 • MW: 39.6 kDa