Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human FGFR3 [G697C], GST Tag, His Tag Recombinant Protein

Catalog No. PV6186
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV6186 1 mg
PV6185 100 μg
PV6184 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PV6186 Supplier Invitrogen™ Supplier No. PV6186
Add to Cart
Add to Cart
Request a Quote

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Human FGFR3, C-terminal fragment, amino acids R397-T806 (as in NCBI/Protein entry NP_000133.1) with a G697C mutation, N-terminal GST-HIS6 fusion protein with a 3C cleavage site, expressed in Sf9 insect cells. , Validated in LanthaScreen™ Kinase Binding Assay, Protocols available for simple use and optimized performance, Multiple pack sizes available for convenient use Related Product: LanthaScreen™ Eu Kinase Binding Assay Protein Form: Recombinant

This gene encodes a member of the fibroblast growth factor receptor (FGFR) family, with its amino acid sequence being highly conserved between members and among divergent species. FGFR family members differ from one another in their ligand affinities and tissue distribution. A full-length representative protein would consist of an extracellular region, composed of three immunoglobulin-like domains, a single hydrophobic membrane-spanning segment and a cytoplasmic tyrosine kinase domain. The extracellular portion of the protein interacts with fibroblast growth factors, setting in motion a cascade of downstream signals, ultimately influencing mitogenesis and differentiation. This particular family member binds acidic and basic fibroblast growth hormone and plays a role in bone development and maintenance. Mutations in this gene lead to craniosynostosis and multiple types of skeletal dysplasia. Three alternatively spliced transcript variants that encode different protein isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM HEPES with 5mM DTT, 20% glycerol, 15mM glutathione, 100mM NaCl and no preservative; pH 7.5
Molecular Weight (g/mol) 74 kDa
Name Human FGFR3 [G697C], GST Tag, His Tag
pH Range 7.5
Purification Method Purified
Quantity 1 mg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias ACH, CD333, CEK2, FGFR-3, HSFGFR3EX, JTK4
Common Name FGFR3 (G697C Mutant)
Protein Length aa 397-806
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag, His-tag
Sequence 397-806
Content And Storage Storage Buffer: 50 mM HEPES pH 7.5, 100 mM NaCl, 5 mM DTT, 15 mM reduced glutathione, 20% glycerol

Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws.
Kinase Group Receptor Tyrosine Kinase
Expression System Insect cells
Protein Form Recombinant
Form Liquid
Protein Subtype FGFs (Fibroblast Growth Factors)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.