Learn More
Carnabio Usa Inc FGFR3(K650E)/10UG/CYTOPLASMIC
Catalog No. 507540790
Encompass
FGFR3 [K650E], Cytoplasmic domain, Mutant [K650E], Amino Acid 436-806, NP_000133.1, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0790 Supplier Carnabio Usa Inc Supplier No. 0850110
FGFR3 [K650E], Cytoplasmic domain, Mutant [K650E], Amino Acid 436-806, NP_000133.1, Expressed in Insect (sf21); 10microg