Learn More
Eurofins Discoverx FGFR4, active, 250 µg. (14-583M)
Supplier:  Eurofins Discoverx 14583M
Encompass
N-terminal 6xHis-tagged recombinant, human FGFR4 amino acids 442-755. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9006