Learn More
Eurofins Discoverx PathHunter FGFR4-β-Klotho Bioassay Kit, 2pl. (93-1060Y3-00089)
Catalog No. 502001959
Encompass
The PathHunter FGFR4-β-Klotho Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses FGF19-induced activation of FGFR4-β-Klotho RTK receptors via detection of SH2 domain protein recruitment. The included cell line overexpresses ProLink-tagged FGFR4 and an EA-tagged SH2 domain protein. It also overexpresses an untagged coreceptor, β-Klotho. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.
Catalog No. 50-200-1959 Supplier Eurofins Discoverx Supplier No. 931060Y300089
The PathHunter FGFR4-β-Klotho Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses FGF19-induced activation of FGFR4-β-Klotho RTK receptors via detection of SH2 domain protein recruitment. The included cell line overexpresses ProLink-tagged FGFR4 and an EA-tagged SH2 domain protein. It also overexpresses an untagged coreceptor, β-Klotho. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.