Learn More
Medchemexpress LLC HY-100631 5mg Medchemexpress, FGFR4-IN-1 CAS:1708971-72-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1006315MG
Encompass_Preferred
Medchemexpress, HY-100631 5mg FGFR4-IN-1 CAS:1708971-72-5 FGFR4-IN-1 is a potent inhibiotr of FGFR4 with IC 50 of 0.7 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1905