Learn More
Carnabio Usa Inc FGFR4(N535K)/5UG/CYTOPLASMICF
Supplier:  Carnabio Usa Inc 085245
Encompass
FGFR4 [N535K], Cytoplasmic domain, Mutant [N535K], Amino Acid 460-802(end), NP_002002.3, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0438