Learn More
Carnabio Usa Inc FGFR4(V550L)/10UG/CYTOPLASMIC
Supplier:  Carnabio Usa Inc 0852610
Encompass
FGFR4 [V550L], Cytoplasmic domain, Mutant [V550L], Amino Acid 460-802(end), NP_002002.3, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0795