Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FGI-106 tetrahydrochloride | 1149348-10-6 | 99.7% | 604.49 g·mol⁻¹ | C28H42Cl4N6 | 10MG
SDP

Catalog No. 5000236072
Encompass_Preferred

FGI-106 tetrahydrochloride is the tetrahydrochloride salt of FGI-106, a broad-spectrum antiviral small molecule reported to inhibit viruses such as Ebola, Rift Valley fever, and dengue in vitro. It is supplied as a solid tetrahydrochloride salt for research use.

  • Chemical name: FGI-106 tetrahydrochloride (CAS 1149348-10-6).
  • Molecular formula: C28H42Cl4N6; molecular weight: 604.49 g·mol⁻¹.
  • Purity: 99.7% (as supplied).
  • Physical form: light brown to brown solid (tetrahydrochloride salt).
  • Storage: store solid at 4°C, sealed, away from moisture; in solution, -80°C up to 6 months or -20°C up to 1 month.
  • Packaging: available in multiple milligram-scale packages for research use.

Catalog No. 50-002-36072 Supplier Medchemexpress LLC Supplier No. HY124618A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FGI-106 tetrahydrochloride is the tetrahydrochloride salt of FGI-106, a broad-spectrum antiviral small molecule reported to inhibit viruses such as Ebola, Rift Valley fever, and dengue in vitro. It is supplied as a solid tetrahydrochloride salt for research use.

  • Chemical name: FGI-106 tetrahydrochloride (CAS 1149348-10-6).
  • Molecular formula: C28H42Cl4N6; molecular weight: 604.49 g·mol⁻¹.
  • Purity: 99.7% (as supplied).
  • Physical form: light brown to brown solid (tetrahydrochloride salt).
  • Storage: store solid at 4°C, sealed, away from moisture; in solution, -80°C up to 6 months or -20°C up to 1 month.
  • Packaging: available in multiple milligram-scale packages for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.