Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FGI-106 tetrahydrochloride | 1149348-10-6 | 99.7% | 604.49 g/mol | C28H42Cl4N6 | 5MG
SDP

Catalog No. 5000236429
Encompass_Preferred

FGI-106 tetrahydrochloride is a research-grade, broad-spectrum antiviral inhibitor supplied as the tetrahydrochloride salt for in vitro studies. It has reported activity against several viral pathogens and is intended for laboratory research use only. Prepare stocks in DMSO following solubility guidance and handle per standard laboratory safety procedures.

  • Cas number 1149348-10-6.
  • Molecular formula C28H42Cl4N6; molecular weight 604.49 g/mol.
  • Purity approximately 99.7%.
  • Appearance solid, light brown to brown.
  • Soluble in DMSO with heating, ultrasonication, and pH adjustment; reported solubility 6.67 mg/mL in DMSO.
  • Recommended stock concentrations: 1 mM, 5 mM, and 10 mM in DMSO with mg-to-mL conversion guidance.
  • Store sealed, away from moisture; in solvent: -80°C up to 6 months, -20°C up to 1 month.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-36429 Supplier Medchemexpress LLC Supplier No. HY124618A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FGI-106 tetrahydrochloride is a research-grade, broad-spectrum antiviral inhibitor supplied as the tetrahydrochloride salt for in vitro studies. It has reported activity against several viral pathogens and is intended for laboratory research use only. Prepare stocks in DMSO following solubility guidance and handle per standard laboratory safety procedures.

  • Cas number 1149348-10-6.
  • Molecular formula C28H42Cl4N6; molecular weight 604.49 g/mol.
  • Purity approximately 99.7%.
  • Appearance solid, light brown to brown.
  • Soluble in DMSO with heating, ultrasonication, and pH adjustment; reported solubility 6.67 mg/mL in DMSO.
  • Recommended stock concentrations: 1 mM, 5 mM, and 10 mM in DMSO with mg-to-mL conversion guidance.
  • Store sealed, away from moisture; in solvent: -80°C up to 6 months, -20°C up to 1 month.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.