Learn More
TARGETMOL CHEMICALS INC FGI-106 tetrahydrochloride 5MG
SDP
Catalog No. 502878504
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. FGI-106 tetrahydrochloride demonstrates potent inhibitory activity across a broad spectrum of viruses, including those causing hemorrhagic fevers such as Ebola, Rift Valley, and Dengue Fever, with half-maximal effective concentrations (EC50s) of 100 nM, 800 nM, and 400-900 nM, respectively. Additionally, it effectively inhibits non-hemorrhagic fever viruses, notably HCV and HIV-1, with EC50s of 200 nM and 150 nM, respectively, indicating its broad-spectrum antiviral potential. Purity 98.98%
Catalog No. 50-287-8504 Supplier TARGETMOL CHEMICALS INC Supplier No. T11281L5MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. FGI-106 tetrahydrochloride demonstrates potent inhibitory activity across a broad spectrum of viruses, including those causing hemorrhagic fevers such as Ebola, Rift Valley, and Dengue Fever, with half-maximal effective concentrations (EC50s) of 100 nM, 800 nM, and 400-900 nM, respectively. Additionally, it effectively inhibits non-hemorrhagic fever viruses, notably HCV and HIV-1, with EC50s of 200 nM and 150 nM, respectively, indicating its broad-spectrum antiviral potential. Purity 98.98%