Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fgin 1-27 | 142720-24-9 | 100.0% | 436.60 | C28H37FN2O | 10 MG
SDP

Catalog No. 5000211220
Encompass_Preferred

FGIN 1-27 is a selective peripheral benzodiazepine receptor ligand supplied as a high-purity research compound for in vitro and in vivo neuroscience studies. It is provided as a solid and in ready-to-use solution formats, with documented solubility and storage guidance to support experimental workflows.

  • High purity suitable for receptor binding and pharmacology assays.
  • Solid and solution formats for flexible dosing and formulation.
  • Soluble in DMSO; in vivo formulation protocols available.
  • Storage conditions provided for long-term and short-term stability.
  • Datasheet, certificate of analysis, and safety data available for experimental documentation.

Catalog No. 50-002-11220 Supplier Medchemexpress LLC Supplier No. HY10105910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FGIN 1-27 is a selective peripheral benzodiazepine receptor ligand supplied as a high-purity research compound for in vitro and in vivo neuroscience studies. It is provided as a solid and in ready-to-use solution formats, with documented solubility and storage guidance to support experimental workflows.

  • High purity suitable for receptor binding and pharmacology assays.
  • Solid and solution formats for flexible dosing and formulation.
  • Soluble in DMSO; in vivo formulation protocols available.
  • Storage conditions provided for long-term and short-term stability.
  • Datasheet, certificate of analysis, and safety data available for experimental documentation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.