Learn More
Medchemexpress LLC Fgin 1-27 | 142720-24-9 | 100.0% | 436.60 | C28H37FN2O | 50 MG
SDP
Catalog No. 5000212155
Encompass_Preferred
FGIN 1-27 is a small-molecule ligand for the translocator protein (TSPO/PBR) used in preclinical research to study mitochondrial benzodiazepine receptor function and neuropharmacology. It is supplied as a solid research chemical with a defined purity and storage recommendations suitable for laboratory assays.
  • High purity for reproducible experimental results.
  • Solid, off-white to light yellow material for easy handling.
  • Storage recommendations support long-term stability at low temperatures.
  • Available in small pack sizes for flexibility in dosing and experiments.
  • Well-characterized chemical identifiers (CAS 142720-24-9) for unambiguous identification.
Catalog No. 50-002-12155 Supplier Medchemexpress LLC Supplier No. HY10105950MG
FGIN 1-27 is a small-molecule ligand for the translocator protein (TSPO/PBR) used in preclinical research to study mitochondrial benzodiazepine receptor function and neuropharmacology. It is supplied as a solid research chemical with a defined purity and storage recommendations suitable for laboratory assays.
  • High purity for reproducible experimental results.
  • Solid, off-white to light yellow material for easy handling.
  • Storage recommendations support long-term stability at low temperatures.
  • Available in small pack sizes for flexibility in dosing and experiments.
  • Well-characterized chemical identifiers (CAS 142720-24-9) for unambiguous identification.