Learn More
ACROBiosystems ACROBiosystems Biotinylated Human FGL1 Protein, Avitag™,Fc Tag
Catalog No. 501956636
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human FGL1, Avitag,Fc Tag (FG1-H82F4) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Ile 312 (Accession # Q08830-1).;This protein carries an Avi tag (Avitag™) at the N-terminus, followed by a human IgG1 Fc tag. The protein has a calculated MW of 62.1 kDa. The protein migrates as 65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human LAG-3, Llama IgG2b Fc Tag, low endotoxin (Cat. No. LA3-H525c) at 5 μg/mL (100 μL/well) can bind Biotinylated Human FGL1, Avitag,Fc Tag (Cat. No. FG1-H82F4) with a linear range of 0.156-5 μg/mL (QC tested).;FACS analysis shows that Biotinylated Human FGL1, Avitag,Fc Tag (Cat. No. FG1-H82F4) can bind to 293T cells overexpressing human LAG3. The concentration of Human FGL1 is 10 μg/mL (Routinely tested).
Catalog No. 50-195-6636 Supplier ACROBiosystems Supplier No. FG1H82F4200UG
Recombinant Protein;200UG;Biotinylated Human FGL1, Avitag,Fc Tag (FG1-H82F4) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Ile 312 (Accession # Q08830-1).;This protein carries an Avi tag (Avitag™) at the N-terminus, followed by a human IgG1 Fc tag. The protein has a calculated MW of 62.1 kDa. The protein migrates as 65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human LAG-3, Llama IgG2b Fc Tag, low endotoxin (Cat. No. LA3-H525c) at 5 μg/mL (100 μL/well) can bind Biotinylated Human FGL1, Avitag,Fc Tag (Cat. No. FG1-H82F4) with a linear range of 0.156-5 μg/mL (QC tested).;FACS analysis shows that Biotinylated Human FGL1, Avitag,Fc Tag (Cat. No. FG1-H82F4) can bind to 293T cells overexpressing human LAG3. The concentration of Human FGL1 is 10 μg/mL (Routinely tested).