Learn More
ACROBiosystems ACROBiosystems Cynomolgus / Rhesus macaque FGL1 Protein, Fc Tag
Supplier:  ACROBiosystems FG1C52691MG
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus / Rhesus macaque FGL1, Fc Tag (FG1-C5269) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Ile 312 (Accession # G7N0K6-1).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 60.4 kDa. The protein migrates as 65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human LAG-3, Mouse IgG2a Fc Tag (Cat. No. LA3-H52Aa) at 10 μg/mL (100 μL/well) can bind Cynomolgus / Rhesus macaque FGL1, Fc Tag (Cat. No. FG1-C5269) with a linear range of 0.01-0.313 μg/mL (QC tested).
Catalog No. 50-195-6531