Learn More
AdipoGen FGL1 h Matched Pair mAbs
SDP
Catalog No. 502562727
Encompass_Preferred
Matched Pair Detection Set. Detects human FGL1 in serum, plasma and cell culture supernatant. Assay Type Sandwich. Detection Type Colorimetric. Sample Type Cell Culture Supernatant, Plasma, Serum. Range 7.8 pg/ml to 500 pg/ml. Sensitivity <5 pg/ml. FGL1 Fibrinogen-like protein 1, also called Hepatocyte-derived fibrinogen-related protein 1, HFREP-1 or Hepassocin was initially identified as an overexpressed transcript in hepatocyte carcinoma and as a transcript enriched in regenerating liver. FGL1 is expressed at lower levels in brown and white adipose in the setting of liver injury. A low level expression of FGL1 is also observed in the pancreas. FGL1 is a 34kDa protein structurally similar to Angiopoietin-like factors 2, 3, 4 and 6, which regulate lipid metabolism and energy utilization. It was proposed that FGL1 is a member of an emerging group of proteins having potential roles in liver metabolism and liver regeneration.
Catalog No. 50-256-2727 Supplier AdipoGen Supplier No. AG46B0014KI01
Matched Pair Detection Set. Detects human FGL1 in serum, plasma and cell culture supernatant. Assay Type Sandwich. Detection Type Colorimetric. Sample Type Cell Culture Supernatant, Plasma, Serum. Range 7.8 pg/ml to 500 pg/ml. Sensitivity <5 pg/ml. FGL1 Fibrinogen-like protein 1, also called Hepatocyte-derived fibrinogen-related protein 1, HFREP-1 or Hepassocin was initially identified as an overexpressed transcript in hepatocyte carcinoma and as a transcript enriched in regenerating liver. FGL1 is expressed at lower levels in brown and white adipose in the setting of liver injury. A low level expression of FGL1 is also observed in the pancreas. FGL1 is a 34kDa protein structurally similar to Angiopoietin-like factors 2, 3, 4 and 6, which regulate lipid metabolism and energy utilization. It was proposed that FGL1 is a member of an emerging group of proteins having potential roles in liver metabolism and liver regeneration.