Learn More
ACROBiosystems ACROBiosystems Human FGL1 Protein, His Tag
Catalog No. 501956642
Encompass_Preferred
Recombinant Protein;1MG;Human FGL1, His Tag (FG1-H52Hy) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Ile 312 (Accession # Q08830-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 36.0 kDa. The protein migrates as 33-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;FACS assay shows that Human FGL1, His Tag (Cat. No. FG1-H52Hy) can bind to 293T cells overexpressing human LAG3. The concentration of Human FGL1 is 20 μg/mL (Routinely tested).
Catalog No. 50-195-6642 Supplier ACROBiosystems Supplier No. FG1H52HY1MG
Recombinant Protein;1MG;Human FGL1, His Tag (FG1-H52Hy) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Ile 312 (Accession # Q08830-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 36.0 kDa. The protein migrates as 33-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;FACS assay shows that Human FGL1, His Tag (Cat. No. FG1-H52Hy) can bind to 293T cells overexpressing human LAG3. The concentration of Human FGL1 is 20 μg/mL (Routinely tested).