Learn More
ACROBiosystems ACROBiosystems Cynomolgus / Rhesus macaque FGL1 Protein, Mouse IgG2a Fc Tag
Supplier:  ACROBiosystems FG1C52501MG
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus / Rhesus macaque FGL1, Mouse IgG2a Fc Tag (FG1-C5250) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Ile 312 (Accession # G7N0K6-1).;This protein carries a mouse IgG2a Fc tag at the N-terminus. The protein has a calculated MW of 60.8 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human LAG-3, Fc,Avitag (Cat. No. LA3-H82Fb) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Cynomolgus / Rhesus macaque FGL1, Mouse IgG2a Fc Tag (Cat. No. FG1-C5250) with a linear range of 0.019-0.625 μg/mL (QC tested).
Catalog No. 50-195-6432