Learn More
Genscript Corporation FGL2Biotin His Avi Flag
Catalog No. 5000059253
Encompass
Fibrinogen-like protein 2 (FGL2) is a member of the fibrinogen-like protein family and possesses important regulatory functions in both innate and adaptive immune responses FGL2 is overexpressed in glioma and its expression level is negatively associated with the prognosis of glioma patients
Catalog No. 50-000-59253 Supplier Genscript Corporation Supplier No. Z04465100
Fibrinogen-like protein 2 (FGL2) is a member of the fibrinogen-like protein family and possesses important regulatory functions in both innate and adaptive immune responses FGL2 is overexpressed in glioma and its expression level is negatively associated with the prognosis of glioma patients