Learn More
Genscript Corporation FGL2Biotin His Avi Flag
Supplier:  Genscript Corporation Z04364500
Encompass
Fibrinogen-like protein 2 (FGL2) is a member of the fibrinogen-like protein family and possesses important regulatory functions in both innate and adaptive immune responses FGL2 is overexpressed in glioma and its expression level is negatively associated with the prognosis of glioma patients
Catalog No. 50-000-58591