Learn More
Lonza Walkersville FGM-2 Fibroblast Growth Medium-2 SingleQuots Supplements and Growth Factors
Catalog No. NC9790054
Encompass
FGM -2 SingleQuot Kit are components sold separately and does not include the basal medium. These are part of the FGM -2 Fibroblast Growth Medium BulletKit (Catalog Number CC-3132) which consists of basal medium and supplements packaged together.FGM-2 Fibroblast Growth Medium-2 BulletKit was developed to support the growth of most primary human fibroblasts including lung and dermal fibroblasts. The kit contains basal medium and supplements packaged together for convenience to culture fibroblasts. FGM-2 Medium contains 2% serum.Purchase our Lung Fibroblast Cell Culture Kit (catalog no. CC-2512B) and get a discounted price on cells and media. The kit consists of a cryopreserved ampule of lung fibroblast cells (catalog no. CC-2512) and FGM-2 BulletKit Growth Media (catalog no. CC-3132).Content:1 x Lilac Cap Vial with Insulin 0.50 mL1 x Gray Cap Vial with hFGF-B 0.50 mL1 x Red Cap Vial with GA-1000 0.50 mL1 x Bottle FBS 10.00 mL
Catalog No. NC9790054 Supplier Lonza Walkersville Supplier No. CC4126
FGM -2 SingleQuot Kit are components sold separately and does not include the basal medium. These are part of the FGM -2 Fibroblast Growth Medium BulletKit (Catalog Number CC-3132) which consists of basal medium and supplements packaged together.FGM-2 Fibroblast Growth Medium-2 BulletKit was developed to support the growth of most primary human fibroblasts including lung and dermal fibroblasts. The kit contains basal medium and supplements packaged together for convenience to culture fibroblasts. FGM-2 Medium contains 2% serum.Purchase our Lung Fibroblast Cell Culture Kit (catalog no. CC-2512B) and get a discounted price on cells and media. The kit consists of a cryopreserved ampule of lung fibroblast cells (catalog no. CC-2512) and FGM-2 BulletKit Growth Media (catalog no. CC-3132).Content:1 x Lilac Cap Vial with Insulin 0.50 mL1 x Gray Cap Vial with hFGF-B 0.50 mL1 x Red Cap Vial with GA-1000 0.50 mL1 x Bottle FBS 10.00 mL