Learn More
Lonza Walkersville FGM3 Cardiac Fibroblast Growth Medium-3 SingleQuots Supplements and Growth Factors
Catalog No. NC0989794
Encompass
FGM -3 SingleQuot Kit are components sold separately and does not include the basal medium. These are part of the FGM -3 Fibroblast Growth Medium BulletKit (Catalog Number CC-4526) which consists of basal medium and supplements packaged together.FGM-3 Fibroblast Growth Medium-3 was developed specifically for cardiac fibroblasts. FGM-3 Medium contains contains 10% serum.Content:1 x Lilac Cap Vial with Insulin 0.50 mL1 x Gray Cap Vial with hFGF-B 0.50 mL1 x Red Cap Vial with GA-1000 0.50 mL1 x Bottle FBS 50.00 mL
Catalog No. NC0989794 Supplier Lonza Walkersville Supplier No. CC4525
FGM -3 SingleQuot Kit are components sold separately and does not include the basal medium. These are part of the FGM -3 Fibroblast Growth Medium BulletKit (Catalog Number CC-4526) which consists of basal medium and supplements packaged together.FGM-3 Fibroblast Growth Medium-3 was developed specifically for cardiac fibroblasts. FGM-3 Medium contains contains 10% serum.Content:1 x Lilac Cap Vial with Insulin 0.50 mL1 x Gray Cap Vial with hFGF-B 0.50 mL1 x Red Cap Vial with GA-1000 0.50 mL1 x Bottle FBS 50.00 mL