Learn More
Lonza Walkersville FGM-2 Fibroblast Growth Medium-2 BulletKit
Catalog No. NC9462405
Encompass
FGM-2 Fibroblast Growth Medium-2 was developed to support the growth of most primary human fibroblasts including lung and dermal fibroblasts. The kit contains basal medium and supplements packaged together for convenience to culture fibroblasts. FGM-2 Medium contains 2% serum.Purchase our Lung Fibroblast Cell Culture Kit (catalog no. CC-2512B) and get a discounted price on cells and media. The kit consists of a cryopreserved ampule of lung fibroblast cells (catalog no. CC-2512) and FGM-2 BulletKit Growth Media (catalog no. CC-3132).Content:1 x FBM Basal Medium (CC-3131), 500 mL1 x FGM-2 SingleQuots Supplement Pack (CC-4126) containing:1 x Lilac Cap Vial with Insulin, 0.50 mL1 x Gray Cap Vial with hFGF-B, 0.50 mL1 x Red Cap Vial with GA-1000, 0.50 mL1 x Bottle FBS, 10.00 mL
Catalog No. NC9462405 Supplier Lonza Walkersville Supplier No. CC3132
FGM-2 Fibroblast Growth Medium-2 was developed to support the growth of most primary human fibroblasts including lung and dermal fibroblasts. The kit contains basal medium and supplements packaged together for convenience to culture fibroblasts. FGM-2 Medium contains 2% serum.Purchase our Lung Fibroblast Cell Culture Kit (catalog no. CC-2512B) and get a discounted price on cells and media. The kit consists of a cryopreserved ampule of lung fibroblast cells (catalog no. CC-2512) and FGM-2 BulletKit Growth Media (catalog no. CC-3132).Content:1 x FBM Basal Medium (CC-3131), 500 mL1 x FGM-2 SingleQuots Supplement Pack (CC-4126) containing:1 x Lilac Cap Vial with Insulin, 0.50 mL1 x Gray Cap Vial with hFGF-B, 0.50 mL1 x Red Cap Vial with GA-1000, 0.50 mL1 x Bottle FBS, 10.00 mL