Learn More
Carnabio Usa Inc FGR/5UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 081665
Encompass
FGR, Full-length, Wild type, Amino Acid 1-529, NP_005239.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0441