Learn More
Eurofins Discoverx Fgr, active, 10 µg. (14-568)
Supplier:  Eurofins Discoverx 14568
Encompass
N-terminal 6xHis-tagged recombinant, human Fgr amino acids 2-end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8981