Learn More
Abcam FH-535, Wnt/beta-cantenin inhibitor, 50MG
Catalog No. 502705592
Encompass_Preferred
MW 361.2 Da, Purity >98%. Wnt/β-cantenin inhibitor and PPARγ and PPARδ antagonist. Suppresses proliferation of cancer cells in vitro. Antitumor agent.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5592 Supplier Abcam Supplier No. AB14459450MG
MW 361.2 Da, Purity >98%. Wnt/β-cantenin inhibitor and PPARγ and PPARδ antagonist. Suppresses proliferation of cancer cells in vitro. Antitumor agent.
The product is subject to the following: Abcam Restricted Use Statement