Learn More
Medchemexpress LLC FH1 (NSC 12407) | 2719-05-3 | MFCD00027653 | 99.2% | 282.34 | C17H18N2O2 | 100MG
SDP
Catalog No. 5000239090
Encompass_Preferred
FH1 (NSC 12407) is a small-molecule hepatocyte function enhancer used in cell-based research to promote hepatocyte differentiation and improve liver cell function. Supplied as a solid (yellow to orange), it is characterized by high purity and defined storage conditions for reliable experimental use.
  • Promotes hepatocyte differentiation and maturation in vitro.
  • Modulates quinone reductase (NQO2) pathway relevant to liver function.
  • High purity (99.2%) suitable for biological assays.
  • Chemical formula C17H18N2O2; molecular weight 282.34.
  • Available in multiple package sizes for experimental flexibility.
  • Stable when stored as powder at -20 °C for long-term storage.
Catalog No. 50-002-39090 Supplier Medchemexpress LLC Supplier No. HY12346100MG
Add to Cart
Add to Cart
FH1 (NSC 12407) is a small-molecule hepatocyte function enhancer used in cell-based research to promote hepatocyte differentiation and improve liver cell function. Supplied as a solid (yellow to orange), it is characterized by high purity and defined storage conditions for reliable experimental use.
  • Promotes hepatocyte differentiation and maturation in vitro.
  • Modulates quinone reductase (NQO2) pathway relevant to liver function.
  • High purity (99.2%) suitable for biological assays.
  • Chemical formula C17H18N2O2; molecular weight 282.34.
  • Available in multiple package sizes for experimental flexibility.
  • Stable when stored as powder at -20 °C for long-term storage.